AMBH2D6F5823 Ambeed, Inc.

(S)-tert-Butyl 3-aminoazepane-1-carboxylate

  • MDL number MFCD11041390
  • Cas Number : 625471-04-7
  • Molecular Weight : 214.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Flash Point 133.1
Storage temp. Keep in dark place,Inert atmosphere,2-8
Physical Form liquid
Shipping Temperature Normal
Country of Origin CN
Boiling point 296.5�0�t 760 mmHg
Product Link https://www.ambeed.com/products/625471-04-7.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/625471-04-7.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart