AMBH2D6F583D Ambeed, Inc.

7-Iodo-2H-benzo[b][1,4]oxazin-3(4H)-one

  • MDL number MFCD07774202
  • Cas Number : 874840-87-6
  • Molecular Weight : 275.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 7-iodo-2H-1,4-benzoxazin-3(4H)-one
Inchi Code 1S/C8H6INO2/c9-5-1-2-6-7(3-5)12-4-8(11)10-6/h1-3H,4H2,(H,10,11)
Inchi Key RNGWLYQJCXPBKZ-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Keep in dark place,Sealed in dry,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/874840-87-6.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/874840-87-6.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart