AMBH2D6F5877 Ambeed, Inc.

2-Chloro-N-(pyridin-4-yl)acetamide

  • MDL number MFCD00996250
  • Cas Number : 80650-46-0
  • Molecular Weight : 170.5963

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/80650-46-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/80650-46-0.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart