AMBH2D6F58E8 Ambeed, Inc.

4-Aminoisothiochroman 2,2-dioxide hydrochloride

  • MDL number MFCD11616514
  • Cas Number : 1187830-61-0
  • Molecular Weight : 233.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/1187830-61-0.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/1187830-61-0.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart