AMBH2D6F58FA Ambeed, Inc.

5,6,7,8-Tetrahydro-1,6-naphthyridine hydrochloride

  • MDL number MFCD11846187
  • Cas Number : 1187830-51-8
  • Molecular Weight : 170.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/1187830-51-8.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1187830-51-8.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart