AMBH2D6F5952 Ambeed, Inc.

5-Methyl-1H-pyrrole-3-carboxylic acid

  • MDL number MFCD09991919
  • Cas Number : 100047-52-7
  • Molecular Weight : 125.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 326.3�t 760 mmHg
Iupac Name 5-methyl-1H-pyrrole-3-carboxylic acid
Inchi Code 1S/C6H7NO2/c1-4-2-5(3-7-4)6(8)9/h2-3,7H,1H3,(H,8,9)
Inchi Key COMZOQAWNKVQEZ-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Sealed in dry,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 326.3�t 760 mmHg
Product Link https://www.ambeed.com/products/100047-52-7.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/100047-52-7.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart