AMBH2D6F5966 Ambeed, Inc.

2,3-Dichloroquinoline

  • MDL number MFCD00234477
  • Cas Number : 613-18-3
  • Molecular Weight : 198.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Boiling point 288.7°C at 760 mmHg
Product Link https://www.ambeed.com/products/613-18-3.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/613-18-3.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart