AMBH2D6F5967 Ambeed, Inc.

5-Bromo-1-chloro-3-fluoro-2-iodobenzene

  • MDL number MFCD09878165
  • Cas Number : 83027-73-0
  • Molecular Weight : 335.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/83027-73-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/83027-73-0.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart