AMBH2D6F5A13 Ambeed, Inc.

Methyl 4-fluoro-2,6-dimethylbenzoate

  • MDL number MFCD06203803
  • Cas Number : 14659-60-0
  • Molecular Weight : 182.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 29-30
Purity 97
Flash Point 85.6
Storage temp. Sealed in dry,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 221.7�0�t 760 mmHg
Product Link https://www.ambeed.com/products/14659-60-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/14659-60-0.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart