AMBH2D6F5A18 Ambeed, Inc.

3-Methoxyfuran-2-carbaldehyde

  • MDL number MFCD15143812
  • Cas Number : 32487-58-4
  • Molecular Weight : 126.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/32487-58-4.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/32487-58-4.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart