AMBH2D6F5A85 Ambeed, Inc.

Fmoc-HoArg-OH

  • MDL number MFCD01861360
  • Cas Number : 776277-76-0
  • Molecular Weight : 410.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-6-guanidinohexanoic acid
Purity 95
Storage temp. Keep in dark place,Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/776277-76-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/776277-76-0.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart