AMBH2D6F5AAF Ambeed, Inc.

4,4,5,5-Tetramethyl-2-(2-(naphthalen-2-yl)phenyl)-1,3,2-dioxaborolane

  • MDL number MFCD22421637
  • Cas Number : 1062555-59-2
  • Molecular Weight : 330.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1062555-59-2.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/1062555-59-2.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart