AMBH2D6F682F Ambeed, Inc.

6-Bromo-1,2,3,4-tetrahydronaphthalen-1-amine

  • MDL number MFCD20463916
  • Cas Number : 1337523-99-5
  • Molecular Weight : 226.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Inert atmosphere,2-8
Physical Form Pale-yellow to Yellow-brown to Black Sticky Oil to Semi-Solid or Liquid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/1337523-99-5.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1337523-99-5.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart