AMBH2D6F6839 Ambeed, Inc.

Pyromeconic Acid

  • MDL number MFCD09863957
  • Cas Number : 496-63-9
  • Molecular Weight : 112.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 117-118
Synonym 3-Hydroxy-4H-pyran-4-one
Purity 97
Flash Point 139.9
Storage temp. Sealed in dry,Store in freezer, under -20
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 298.7�t 760 mmHg
Product Link https://www.ambeed.com/products/496-63-9.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/496-63-9.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart