AMBH2D6F6845 Ambeed, Inc.

5-Bromo-1H-pyrrolo[2,3-b]pyridine-2,3-dione

  • MDL number MFCD13193450
  • Cas Number : 149142-67-6
  • Molecular Weight : 227.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/149142-67-6.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/149142-67-6.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart