AMBH2D6F6A50 Ambeed, Inc.

1-(Benzyloxy)-6-oxo-1,6-dihydropyridine-2-carboxylic acid

  • MDL number MFCD20270106
  • Cas Number : 210366-15-7
  • Molecular Weight : 245.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/210366-15-7.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/210366-15-7.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart