AMBH2D6F6AB2 Ambeed, Inc.

2-(3,4-Dihydroxyphenyl)-2-((2-(3,4-dihydroxyphenyl)-5,7-dihydroxychroman-3-yl)oxy)chroman-3,4,5,7-tetraol

  • MDL number MFCD01662851
  • Cas Number : 20347-71-1
  • Molecular Weight : 594.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/20347-71-1.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/20347-71-1.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart