AMBH2D6F6ADF Ambeed, Inc.

Methyl 3-amino-2,4-dimethylbenzoate

  • MDL number MFCD22494937
  • Cas Number : 24812-89-3
  • Molecular Weight : 179.2200

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Keep in dark place,Inert atmosphere,2-8
Physical Form liquid
Shipping Temperature Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/24812-89-3.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/24812-89-3.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart