AMBH2D6F6AE9 Ambeed, Inc.

4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2(1H)-one

  • MDL number MFCD23140955
  • Cas Number : 1256358-90-3
  • Molecular Weight : 221.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-ol
Purity 97
Storage temp. Inert atmosphere,Store in freezer, under -20
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/1256358-90-3.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1256358-90-3.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart