AMBH2D6F6C06 Ambeed, Inc.

6'-(Dibutylamino)-3'-methyl-2'-(phenylamino)-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one

  • MDL number MFCD00307579
  • Cas Number : 89331-94-2
  • Molecular Weight : 532.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/89331-94-2.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/89331-94-2.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart