AMBH2D6F6C52 Ambeed, Inc.

1-Phenyl-1,2-dihydroisoquinolin-3(4H)-one

  • MDL number MFCD00089450
  • Cas Number : 17507-05-0
  • Molecular Weight : 223.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/17507-05-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/17507-05-0.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart