AMBH2D6F6C58 Ambeed, Inc.

(2-Bromo-1-fluoroethyl)benzene

  • MDL number MFCD13689011
  • Cas Number : 1786-36-3
  • Molecular Weight : 203.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Flash Point 82.5
Storage temp. Inert atmosphere,2-8
Physical Form liquid
Shipping Temperature Normal
Country of Origin CN
Boiling point 208.3�0�t 760 mmHg
Product Link https://www.ambeed.com/products/1786-36-3.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1786-36-3.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart