AMBH2D6F6C7C Ambeed, Inc.

Methyl 5-amino-6-methoxynicotinate

  • MDL number MFCD20697606
  • Cas Number : 59237-50-2
  • Molecular Weight : 182.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Flash Point 156.3
Storage temp. Keep in dark place,Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 334.8�0�t 760 mmHg
Product Link https://www.ambeed.com/products/59237-50-2.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/59237-50-2.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart