AMBH2D6F6CC3 Ambeed, Inc.

1-(3-(tert-Butyl)phenyl)ethanone

  • MDL number MFCD11113485
  • Cas Number : 6136-71-6
  • Molecular Weight : 176.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/6136-71-6.html

Safety Information

signalword Warning
pcodes P280
MSDS https://www.ambeed.com/products/6136-71-6.html
pictograms GHS07
hstatements H227-H302
Availability details for
Quantity
  • Availability
  • Add to Cart