AMBH2D6F6D5C Ambeed, Inc.

(S)-Methyl 2-(pyrrolidin-2-yl)acetate hydrochloride

  • MDL number MFCD22200809
  • Cas Number : 259868-83-2
  • Molecular Weight : 179.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/259868-83-2.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/259868-83-2.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart