AMBH2D6F6DAF Ambeed, Inc.

4-(6-((Methylsulfonyl)oxy)benzo[b]thiophen-2-yl)phenyl methanesulfonate

  • MDL number MFCD16620994
  • Cas Number : 84449-65-0
  • Molecular Weight : 398.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/84449-65-0.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/84449-65-0.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart