AMBH2D6F6DD0 Ambeed, Inc.

H-Tyr-OBzl

  • MDL number MFCD00191034
  • Cas Number : 42406-77-9
  • Molecular Weight : 271.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Benzyl L-tyrosinate
Purity 95
Flash Point 219.1
Storage temp. Keep in dark place,Inert atmosphere,Store in freezer, under -20
Physical Form solid
Shipping Temperature Shipped in cold pack
Country of Origin CN
Boiling point 438.7�t 760 mmHg
Product Link https://www.ambeed.com/products/42406-77-9.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/42406-77-9.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart