AMBH2D6F6E2F Ambeed, Inc.

2-Chlorothiazole-4-carbonitrile

  • MDL number MFCD13179597
  • Cas Number : 944124-72-5
  • Molecular Weight : 144.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/944124-72-5.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/944124-72-5.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart