AMBH2D6F6E6B Ambeed, Inc.

(11bS)-4-Hydroxy-2,6-diphenyldinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide

  • MDL number MFCD17676194
  • Cas Number : 874948-59-1
  • Molecular Weight : 500.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/874948-59-1.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/874948-59-1.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart