AMBH2D6F6E76 Ambeed, Inc.

Ondansetron

  • MDL number MFCD00833882
  • Cas Number : 99614-02-5
  • Molecular Weight : 293.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/99614-02-5.html

Safety Information

signalword Danger
pcodes P301+P310
MSDS https://www.ambeed.com/products/99614-02-5.html
pictograms GHS06
hstatements H301
Availability details for
Quantity
  • Availability
  • Add to Cart