AMBH2D6F6EEB Ambeed, Inc.

Methyl 2-(triphenylphosphoranylidene)propanoate

  • MDL number MFCD08062441
  • Cas Number : 2605-68-7
  • Molecular Weight : 348.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 153-156
Purity 95
Flash Point 260.8
Storage temp. Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 485.8�0�t 760 mmHg
Product Link https://www.ambeed.com/products/2605-68-7.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/2605-68-7.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart