AMBH2D6F6EED Ambeed, Inc.

N-(3-Acetyl-2-hydroxyphenyl)-4-(4-phenylbutoxy)benzamide

  • MDL number MFCD09835307
  • Cas Number : 136450-06-1
  • Molecular Weight : 403.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/136450-06-1.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/136450-06-1.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart