AMBH2D6F6FDA Ambeed, Inc.

4-Methoxy-1H-pyrazolo[3,4-b]pyridine

  • MDL number MFCD11846316
  • Cas Number : 119368-03-5
  • Molecular Weight : 149.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/119368-03-5.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/119368-03-5.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart