AMBH2D6F707F Ambeed, Inc.

(3-Methacrylamidophenyl)boronic acid

  • MDL number MFCD09475805
  • Cas Number : 48150-45-4
  • Molecular Weight : 205.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/48150-45-4.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/48150-45-4.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart