AMBH2D6F70CC Ambeed, Inc.

3-Tosyl-3-azabicyclo[3.2.0]heptan-6-one

  • MDL number MFCD12911657
  • Cas Number : 122080-99-3
  • Molecular Weight : 265.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Store in freezer, under -20°C
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Boiling point 437.5°55.0°C at 760 mmHg
Product Link https://www.ambeed.com/products/122080-99-3.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/122080-99-3.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart