AMBH2D6F70CE Ambeed, Inc.

2-Chloroquinolin-3-ol

  • MDL number MFCD11040214
  • Cas Number : 128676-94-8
  • Molecular Weight : 179.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point >210
Purity 98
Storage temp. Inert atmosphere,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 315.2�t 760 mmHg
Product Link https://www.ambeed.com/products/128676-94-8.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/128676-94-8.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart