AMBH2D6F7140 Ambeed, Inc.

3-Methyl-1H-imidazo[4,5-b]pyridin-2(3H)-one

  • MDL number MFCD01081498
  • Cas Number : 21991-39-9
  • Molecular Weight : 149.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/21991-39-9.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/21991-39-9.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart