AMBH2D6F719C Ambeed, Inc.

N-(3-(4,5-Dihydrothiazol-2-yl)thiazolidin-2-ylidene)-5H-dibenzo[a,d][7]annulen-5-amine

  • MDL number MFCD17676143
  • Cas Number : 1072145-33-5
  • Molecular Weight : 377.53

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1072145-33-5.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/1072145-33-5.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart