AMBH2D6F71B4 Ambeed, Inc.

4,6-Dibromopyridazin-3-amine

  • MDL number MFCD13689240
  • Cas Number : 1206487-35-5
  • Molecular Weight : 252.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Flash Point 181.5
Storage temp. Keep in dark place,Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 376.6�0�t 760 mmHg
Product Link https://www.ambeed.com/products/1206487-35-5.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/1206487-35-5.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart