AMBH2D6F71CC Ambeed, Inc.

Azulen-2-amine

  • MDL number MFCD18072913
  • Cas Number : 50472-20-3
  • Molecular Weight : 143.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 93-94
Purity 97
Flash Point 157
Storage temp. Keep in dark place,Inert atmosphere,Room temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 306.7�0�t 760 mmHg
Product Link https://www.ambeed.com/products/50472-20-3.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/50472-20-3.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart