AMBH2D6F71E2 Ambeed, Inc.

2-Allyl-1-(6-(2-hydroxypropan-2-yl)pyridin-2-yl)-6-(methylthio)-1H-pyrazolo[3,4-d]pyrimidin-3(2H)-one

  • MDL number MFCD19443207
  • Cas Number : 955369-56-9
  • Molecular Weight : 357.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/955369-56-9.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/955369-56-9.html
pictograms GHS07
hstatements H302-H315-H319
Availability details for
Quantity
  • Availability
  • Add to Cart