AMBH2D6F7264 Ambeed, Inc.

Pyrido[3,2-d]pyrimidin-4(3H)-one

  • MDL number MFCD02645992
  • Cas Number : 37538-67-3
  • Molecular Weight : 147.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/37538-67-3.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/37538-67-3.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart