AMBH2D6F7280 Ambeed, Inc.

N-Isopropylbutyramide

  • MDL number MFCD01349261
  • Cas Number : 122348-67-8
  • Molecular Weight : 129.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/122348-67-8.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/122348-67-8.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart