AMBH2D6F728B Ambeed, Inc.

(5-Bromothiophen-2-yl)methanamine hydrochloride

  • MDL number MFCD08752581
  • Cas Number : 1001414-56-7
  • Molecular Weight : 228.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Inert atmosphere,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Product Link https://www.ambeed.com/products/1001414-56-7.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1001414-56-7.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart