AMBH2D6F730B Ambeed, Inc.

5-Bromo-3-methoxypyridin-2(1H)-one

  • MDL number MFCD17676138
  • Cas Number : 1189757-62-7
  • Molecular Weight : 204.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1189757-62-7.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1189757-62-7.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart