AMBH2D6F7325 Ambeed, Inc.

Methyl 5-nitro-1H-indole-3-carboxylate

  • MDL number MFCD06204072
  • Cas Number : 686747-51-3
  • Molecular Weight : 220.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/686747-51-3.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338-P310
MSDS https://www.ambeed.com/products/686747-51-3.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart