AMBH2D6F7346 Ambeed, Inc.

Pyridine-3,5-dicarboxamide

  • MDL number MFCD00229426
  • Cas Number : 4663-99-4
  • Molecular Weight : 165.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/4663-99-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/4663-99-4.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart