AMBH2D6F7350 Ambeed, Inc.

2-Amino-2-(1H-indol-5-yl)acetic acid

  • MDL number MFCD06656882
  • Cas Number : 108763-43-5
  • Molecular Weight : 190.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/108763-43-5.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/108763-43-5.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart