AMBH2D6F736A Ambeed, Inc.

Methyl 3-amino-4-nitrobenzoate

  • MDL number MFCD00017008
  • Cas Number : 99512-09-1
  • Molecular Weight : 196.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Flash Point 192.2°C
Storage temp. Keep in dark place,Inert atmosphere,Room temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Boiling point 394.1°C at 760 mmHg
Product Link https://www.ambeed.com/products/99512-09-1.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/99512-09-1.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart