AMBH2D6F7D7C Ambeed, Inc.

3-Bromo-6-nitroquinoline

  • MDL number MFCD12024480
  • Cas Number : 7101-95-3
  • Molecular Weight : 253.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/7101-95-3.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/7101-95-3.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart